missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HPV HPV16 E6 Protein
A cDNA sequence encoding the HPV16 E6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12310-500ug
Additional Details : Weight : 0.01000kg
Specifications
HPV HPV16 E6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Recombinant HPV16 E6 solution in PBS and 4M Urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HPV | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |