missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HPV HPV16 E6 Protein
A cDNA sequence encoding the HPV16 E6 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | HPV HPV16 E6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HPV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980009
|
enQuireBio™
QP12310-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15900019
|
enQuireBio™
QP12310-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15990009
|
enQuireBio™
QP12310-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HPV HPV16 E6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HPV | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Recombinant HPV16 E6 solution in PBS and 4M Urea. |