missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACP1 / LMW-PTP Protein
A DNA sequence encoding the Homo sapiens (Human) ACP1 / LMW-PTP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5608-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004291.1 | |
ACP1 / LMW-PTP Protein | |
Research Use Only | |
ACP1 | |
Recombinant Protein | |
E. coli | |
MAEQATKSVL FVCLGNICRS PIAEAVFRKL VTDQNISENW RVDSAATSGY EIGNPPDYRG QSCMKRHGIP MSHVARQITK EDFATFDYIL CMDESNLRDL NRKSNQVKTC KAKIELLGSY DPQKQLIIED PYYGNDSDFE TVYQQCVRCC RAFLEKAH | |
Greater than 80% as determined by SDS-PAGE. |
52 | |
1 mg | |
Not determined. | |
Full Length of Isoform 1 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |