missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADSL Protein

A cDNA sequence encoding the ADSL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10973-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955738

  • 203.00 € / 20µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ADSL Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAAG GDHGSPDSYR SPLASRYASP EMCFVFSDRY KFRTWRQLWL WLAEAEQTLG LPITDEQIQE MKSNLENIDF KMAAEEEKRL RHDVMAHVHT FGHCCPKAAG IIHLGATSCY VGDNTDLIIL RNALDLLLPK LARVISRLAD FAKERASLPT LGFTHFQPAQ LTTVGKRCCL WIQDLCMDLQ NLKRVRDDLR FRGVKGTTGT QASFLQLFEG DDHKVEQLDK MVTEKAGFKR AFIITGQTYT RKVDIEVLSV LASLGASVHK ICTDIRLLAN LKEMEEPFEK QQIGSSAMPY KRNPMRSERC CSLARHLMTL VMDPLQTASV QWFERTLDDS ANRRICLAEA FLTADTILNT LQNISEGLVV YPKVIERRIR QELPFMATEN IIMAMVKAGG SRQDCHEKIR VLSQQAASVV KQEGGDNDLI ERIQVDAYFS PIHSQLDHLL DPSSFTGRAS QQVQRFLEEE VYPLLKPYES VMKVKAELCL
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ADSL solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers