missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADSL Protein
A cDNA sequence encoding the ADSL was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | ADSL Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965738
|
enQuireBio™
QP10973-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955738
|
enQuireBio™
QP10973-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945738
|
enQuireBio™
QP10973-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ADSL Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ADSL solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAAG GDHGSPDSYR SPLASRYASP EMCFVFSDRY KFRTWRQLWL WLAEAEQTLG LPITDEQIQE MKSNLENIDF KMAAEEEKRL RHDVMAHVHT FGHCCPKAAG IIHLGATSCY VGDNTDLIIL RNALDLLLPK LARVISRLAD FAKERASLPT LGFTHFQPAQ LTTVGKRCCL WIQDLCMDLQ NLKRVRDDLR FRGVKGTTGT QASFLQLFEG DDHKVEQLDK MVTEKAGFKR AFIITGQTYT RKVDIEVLSV LASLGASVHK ICTDIRLLAN LKEMEEPFEK QQIGSSAMPY KRNPMRSERC CSLARHLMTL VMDPLQTASV QWFERTLDDS ANRRICLAEA FLTADTILNT LQNISEGLVV YPKVIERRIR QELPFMATEN IIMAMVKAGG SRQDCHEKIR VLSQQAASVV KQEGGDNDLI ERIQVDAYFS PIHSQLDHLL DPSSFTGRAS QQVQRFLEEE VYPLLKPYES VMKVKAELCL | |
Greater than 95.0% as determined by SDS-PAGE. |