missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AMD1 Protein

A cDNA sequence encoding the AMD1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11010-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955858

  • 131.00 € / 1µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AMD1 Protein
Research Use Only
Recombinant Protein
E. coli
GSSHHHHHH SSGLVPRGSH MGSHMSSMFV SKRRFILKTC GTTLLLKALV PLLKLARDYS GFDSIQSFFY SRKNFMKPSH QGYPHRNFQE EIEFLNAIFP NGAAYCMGRM NSDCWYLYTL DFPESRVISQ PDQTLEILMS ELDPAVMDQF YMKDGVTAKD VTRESGIRDL IPGSVIDATM FNPCGYSMNG MKSDGTYWTI HITPEPEFSY VSFETNLSQT SYDDLIRKVV EVFKPGKFVT TLFVNQSSKC RTVLASPQKI EGFKRLDCQS AMFNDYNFVF TSFAKKQQQQ QS
Greater than 80% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The AMD1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers