missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AMD1 Protein
A cDNA sequence encoding the AMD1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | AMD1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955858
|
enQuireBio™
QP11010-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975858
|
enQuireBio™
QP11010-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965858
|
enQuireBio™
QP11010-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
AMD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The AMD1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GSSHHHHHH SSGLVPRGSH MGSHMSSMFV SKRRFILKTC GTTLLLKALV PLLKLARDYS GFDSIQSFFY SRKNFMKPSH QGYPHRNFQE EIEFLNAIFP NGAAYCMGRM NSDCWYLYTL DFPESRVISQ PDQTLEILMS ELDPAVMDQF YMKDGVTAKD VTRESGIRDL IPGSVIDATM FNPCGYSMNG MKSDGTYWTI HITPEPEFSY VSFETNLSQT SYDDLIRKVV EVFKPGKFVT TLFVNQSSKC RTVLASPQKI EGFKRLDCQS AMFNDYNFVF TSFAKKQQQQ QS | |
Greater than 80% as determined by SDS-PAGE. |