missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AMD1 Protein
A cDNA sequence encoding the AMD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11010-5ug
Additional Details : Weight : 0.01000kg
Specifications
AMD1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GSSHHHHHH SSGLVPRGSH MGSHMSSMFV SKRRFILKTC GTTLLLKALV PLLKLARDYS GFDSIQSFFY SRKNFMKPSH QGYPHRNFQE EIEFLNAIFP NGAAYCMGRM NSDCWYLYTL DFPESRVISQ PDQTLEILMS ELDPAVMDQF YMKDGVTAKD VTRESGIRDL IPGSVIDATM FNPCGYSMNG MKSDGTYWTI HITPEPEFSY VSFETNLSQT SYDDLIRKVV EVFKPGKFVT TLFVNQSSKC RTVLASPQKI EGFKRLDCQS AMFNDYNFVF TSFAKKQQQQ QS | |
Greater than 80% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The AMD1 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |