missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apolipoprotein C-IV Protein

A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-IV, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8992-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037421

  • 2625.00 € / 500µg
Estimated Shipment: 03-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001637.1
Apolipoprotein C-IV Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
346
Greater than 90% as determined by SDS-PAGE.
Research Use Only
APOC4
Recombinant Protein
Yeast
CQPEAQEGTL SPPPKLKMSR WSLVRGRMKE LLETVVNRTR DGWQWFWSPS TFRGFMQTYY DDHLRDLGPL TKAWFLESKD SLLKKTHSLC PRLVCGDKDQ G
Videos
SDS
Documents
Certificates
Special Offers

Special Offers