missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis-enhancing nuclease Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis-enhancing nuclease, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_073604.3 |
---|---|
Gene ID (Entrez) | 64782 |
Name | Apoptosis-enhancing nuclease Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051051
|
enQuireBio™
QP7830-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071051
|
enQuireBio™
QP7830-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061051
|
enQuireBio™
QP7830-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081051
|
enQuireBio™
QP7830-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091051
|
enQuireBio™
QP7830-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001061
|
enQuireBio™
QP7830-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_073604.3 | |
Apoptosis-enhancing nuclease Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
64782 | |
Research Use Only | |
AEN | |
Recombinant Protein | |
E. coli | |
MVGTGPRGRV SELARCSIVS YHGNVLYDKY IRPEMPIADY RTRWSGITRQ HMRKAVPFQV AQKEILKLLK GKVVVGHALH NDFQALKYVH PRSQTRDTTY VPNFLSEPGL HTRARVSLKD LALQLLHKKI QVGQHGHSSV EDATTAMELY RLVEVQWEQQ EARSLWTCPE DREPDSSTDM EQYMEDQYWP DDLAHGSRGG AREAQDRRN | |
Greater than 90% as determined by SDS-PAGE. |