missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aspartyl / asparaginyl beta-hydroxylase Protein
A DNA sequence encoding the Homo sapiens (Human) Aspartyl/asparaginyl beta-hydroxylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5686-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001158222.1 | |
Aspartyl / asparaginyl beta-hydroxylase Protein | |
Research Use Only | |
ASPH | |
Recombinant Protein | |
E. coli | |
FDLVDYEEVL GKLGIYDADG DGDFDVDDAK VLLGLKERST SEPAVPPEEA EPHTEPEEQV PVEAEPQNIE DEAKEQIQSL LHEMVHAEHE TEHSYHVEET VSQDCNQDME EMMSEQENPD SSEPVVEDER LHHDTDDVTY QVYEEQAVYE PLENEGIEIT EVTAPPEDNP VEDSQVIVEE VSIFPVEEQQ EVPPDT | |
Greater than 90% as determined by SDS-PAGE. |
444 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 6 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |