missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aspartyl / asparaginyl beta-hydroxylase Protein
A DNA sequence encoding the Homo sapiens (Human) Aspartyl/asparaginyl beta-hydroxylase, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001158222.1 |
---|---|
Gene ID (Entrez) | 444 |
Name | Aspartyl / asparaginyl beta-hydroxylase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907009
|
enQuireBio™
QP5686-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927009
|
enQuireBio™
QP5686-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917009
|
enQuireBio™
QP5686-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15937009
|
enQuireBio™
QP5686-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15947009
|
enQuireBio™
QP5686-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957009
|
enQuireBio™
QP5686-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifica
NP_001158222.1 | |
Aspartyl / asparaginyl beta-hydroxylase Protein | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 6 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
444 | |
Research Use Only | |
ASPH | |
Recombinant Protein | |
E. coli | |
FDLVDYEEVL GKLGIYDADG DGDFDVDDAK VLLGLKERST SEPAVPPEEA EPHTEPEEQV PVEAEPQNIE DEAKEQIQSL LHEMVHAEHE TEHSYHVEET VSQDCNQDME EMMSEQENPD SSEPVVEDER LHHDTDDVTY QVYEEQAVYE PLENEGIEIT EVTAPPEDNP VEDSQVIVEE VSIFPVEEQQ EVPPDT | |
Greater than 90% as determined by SDS-PAGE. |