missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Aspartyl / asparaginyl beta-hydroxylase Protein

A DNA sequence encoding the Homo sapiens (Human) Aspartyl/asparaginyl beta-hydroxylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5686-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15907009

  • 505.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001158222.1
Aspartyl / asparaginyl beta-hydroxylase Protein
Research Use Only
ASPH
Recombinant Protein
E. coli
FDLVDYEEVL GKLGIYDADG DGDFDVDDAK VLLGLKERST SEPAVPPEEA EPHTEPEEQV PVEAEPQNIE DEAKEQIQSL LHEMVHAEHE TEHSYHVEET VSQDCNQDME EMMSEQENPD SSEPVVEDER LHHDTDDVTY QVYEEQAVYE PLENEGIEIT EVTAPPEDNP VEDSQVIVEE VSIFPVEEQQ EVPPDT
Greater than 90% as determined by SDS-PAGE.
444
100 μg
Not determined.
Partial (see sequence information for more details). of Isoform 6
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers