missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6432-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033270

  • 518.00 € / 100µg
Estimated Shipment: 03-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003986.2
Atrial natriuretic peptide receptor 2 Protein
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
4882
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NPR2
Recombinant Protein
E. coli
RNLTLAVVLP EHNLSYAWAW PRVGPAVALA VEALGRALPV DLRFVSSELE GACSEYLAPL SAVDLKLYHD PDLLLGPGCV YPAASVARFA SHWRLPLLTA GAVASGFSAK NDHYRTLVRT GPSAPKLGEF VVTLHGHFNW TARAALLYLD ARTDDRPHYF TIEGVFEALQ GSNLSVQHQV YAREPGGPEQ ATHFIRANGR IVYICGPLEM LHEILLQAQR ENLTNGDYVF FYLDVFGESL RAGPTRATGR PWQDNRTREQ AQALREAFQT VLVITYREPP NPEYQEFQNR LLIRAREDFG VELGPSLMNL IAGCFYDGIL LYAEVLNETI QEGGTREDGL RIVEKMQGRR YHGVTGLVVM DKNNDRETDF VLWAMGDLDS GDFQPAAHYS GAEKQIWWTG RPIPWVKGAP PSDNPPCAFD LDDPSCDKTP LSTLAI
Videos
SDS
Documents
Certificates
Special Offers

Special Offers