missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAG family molecular chaperone regulator 2 Protein
A DNA sequence encoding the Homo sapiens (Human) BAG family molecular chaperone regulator 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5703-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004273.1 | |
BAG family molecular chaperone regulator 2 Protein | |
Research Use Only | |
BAG2 | |
Recombinant Protein | |
E. coli | |
AQAKINAKAN EGRFCRSSSM ADRSSRLLES LDQLELRVEA LREAATAVEQ EKEILLEMIH SIQNSQDMRQ ISDGEREELN LTANRLMGRT LTVEVSVETI RNPQQQESLK HATRIIDEVV NKFLDDLGNA KSHLMSLYSA CSSEVPHGPV DQKFQSIVIG CALEDQKKIK RRLETLLRNI ENSDKAIKLL EHSKGAGSKT LQQNAESRFN | |
Greater than 80% as determined by SDS-PAGE. |
9532 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |