missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BP-1 Protein
A cDNA sequence encoding the BP-1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11197-10ug
Additional Details : Weight : 0.01000kg
Specifications
1748 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MTSLPCPLPG RDASKAVFPD LAPVPSVAAA YPLGLSPTTA ASPNLSYSRP YGHLLSYPYT EPANPGDSYL SCQQPAALSQ PLCGPAEHPQ ELEADSEKPR LSPEPSERRP QAPAKKLRKP RTIYSSLQLQ HLNQRFQHTQ YLALPERAQL AAQLGLTQTQ VKIWFQNKRS KYKKLLKQNS GGQEGDFPGR TFSVSPCSPP LPSLWDLPKA GTLPTSGYGN SFGAWYQHHS SDVLASPQMM |
BP-1 Protein | |
Research Use Only | |
DLX4 | |
Human | |
Untagged | |
BP-1 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |