missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BRISC complex subunit Abro1 Protein

A DNA sequence encoding the Homo sapiens (Human) BRISC complex subunit Abro1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7572-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049480

  • 505.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_115558.3
BRISC complex subunit Abro1 Protein
Research Use Only
FAM175B
Recombinant Protein
E. coli
MAASISGYTF SAVCFHSANS NADHEGFLLG EVRQEETFSI SDSQISNTEF LQVIEIHNHQ PCSKLFSFYD YASKVNEESL DRILKDRRKK VIGWYRFRRN TQQQMSYREQ VLHKQLTRIL GVPDLVFLLF SFISTANNST HALEYVLFRP NRRYNQRISL AIPNLGNTSQ QEYKVSSVPN TSQSYAKVIK EHGTDFFDKD GVMKDIRAIY QVYNALQEKV QAVCADVEKS ERVVESCQAE VNKLRRQITQ RKNEKEQERR LQQAVLSRQM PSESLDPAFS PRMPSSGFAA EGRSTLGDAE ASDPPPPYSD FHPNNQESTL SHSRMERSVF MPRPQAVGSS NYASTSAGLK YPGSGADLPP PQRAAGDSGE DSDDSDYENL IDPTEPSNSE YSHSKDSRPM AHPDEDPRNT QTSQI
Greater than 90% as determined by SDS-PAGE.
23172
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers