missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRISC complex subunit Abro1 Protein
A DNA sequence encoding the Homo sapiens (Human) BRISC complex subunit Abro1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_115558.3 |
---|---|
Gene ID (Entrez) | 23172 |
Name | BRISC complex subunit Abro1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049480
|
enQuireBio™
QP7572-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069480
|
enQuireBio™
QP7572-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059480
|
enQuireBio™
QP7572-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079480
|
enQuireBio™
QP7572-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089480
|
enQuireBio™
QP7572-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099480
|
enQuireBio™
QP7572-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_115558.3 | |
BRISC complex subunit Abro1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
23172 | |
Research Use Only | |
FAM175B | |
Recombinant Protein | |
E. coli | |
MAASISGYTF SAVCFHSANS NADHEGFLLG EVRQEETFSI SDSQISNTEF LQVIEIHNHQ PCSKLFSFYD YASKVNEESL DRILKDRRKK VIGWYRFRRN TQQQMSYREQ VLHKQLTRIL GVPDLVFLLF SFISTANNST HALEYVLFRP NRRYNQRISL AIPNLGNTSQ QEYKVSSVPN TSQSYAKVIK EHGTDFFDKD GVMKDIRAIY QVYNALQEKV QAVCADVEKS ERVVESCQAE VNKLRRQITQ RKNEKEQERR LQQAVLSRQM PSESLDPAFS PRMPSSGFAA EGRSTLGDAE ASDPPPPYSD FHPNNQESTL SHSRMERSVF MPRPQAVGSS NYASTSAGLK YPGSGADLPP PQRAAGDSGE DSDDSDYENL IDPTEPSNSE YSHSKDSRPM AHPDEDPRNT QTSQI | |
Greater than 90% as determined by SDS-PAGE. |