missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C1D Protein

A DNA sequence encoding the Homo sapiens (Human) C1D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP938-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038211

  • 324.00 € / 50µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001177192.1
C1D Protein
Research Use Only
C1D
Recombinant Protein
E. coli
MAGEEINEDY PVEIHEYLSA FENSIGAVDE MLKTMMSVSR NELLQKLDPL EQAKVDLVSA YTLNSMFWVY LATQGVNPKE HPVKQELERI RVYMNRVKEI TDKKKAGKLD RGAASRFVKN ALWEPKSKNA SKVAN
Greater than 90% as determined by SDS-PAGE.
10438
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers