missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAB39 Protein
A cDNA sequence encoding the CAB39 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11249-2ug
Additional Details : Weight : 0.01000kg
Specifications
CAB39 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPFPFGKSHK SPADIVKNLK ESMAVLEKQD ISDKKAEKAT EEVSKNLVAM KEILYGTNEK EPQTEAVAQL AQELYNSGLL STLVADLQLI DFEGKKDVAQ IFNNILRRQI GTRTPTVEYI CTQQNILFML LKGYESPEIA LNCGIMLREC IRHEPLAKII LWSEQFYDFF RYVEMSTFDI ASDAFATFKD LLTRHKLLSA EFLEQHYDRF FSEYEKLLHS ENYVTKRQSL KLLGELLLDR HNFTIMTKYISKPENLKLMM NLLRDKSRNI QFEAFHVFKV FVANPNKTQP ILDILLKNQA KLIEFLSKFQ NDRTEDEQFN DEKTYLVKQI RDLKRPAQQE A | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CAB39 solution (0.5 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |