missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAB39 Protein
A cDNA sequence encoding the CAB39 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | CAB39 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15996608
|
enQuireBio™
QP11249-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976608
|
enQuireBio™
QP11249-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986608
|
enQuireBio™
QP11249-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CAB39 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CAB39 solution (0.5 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPFPFGKSHK SPADIVKNLK ESMAVLEKQD ISDKKAEKAT EEVSKNLVAM KEILYGTNEK EPQTEAVAQL AQELYNSGLL STLVADLQLI DFEGKKDVAQ IFNNILRRQI GTRTPTVEYI CTQQNILFML LKGYESPEIA LNCGIMLREC IRHEPLAKII LWSEQFYDFF RYVEMSTFDI ASDAFATFKD LLTRHKLLSA EFLEQHYDRF FSEYEKLLHS ENYVTKRQSL KLLGELLLDR HNFTIMTKYISKPENLKLMM NLLRDKSRNI QFEAFHVFKV FVANPNKTQP ILDILLKNQA KLIEFLSKFQ NDRTEDEQFN DEKTYLVKQI RDLKRPAQQE A | |
Greater than 90.0% as determined by SDS-PAGE. |