missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cadherin-12 Protein

A DNA sequence encoding the Homo sapiens (Human) Cadherin-12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8756-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046311

  • 2285.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001304156.1
Cadherin-12 Protein
Research Use Only
CDH12
Recombinant Protein
E. coli
WVWNQFFVLE EYVGSEPQYV GKLHSDLDKG EGTVKYTLSG DGAGTVFTID ETTGDIHAIR SLDREEKPFY TLRAQAVDIE TRKPLEPESE FIIKVQDIND NEPKFLDGPY VATVPEMSPV GAYVLQVKAT DADDPTYGNS ARVVYSILQG QPYFSIDPKT GVIRTALPNM DREVKEQYQV LIQAKDMGGQ LGGLAGTTIV NITLTDVNDN PPRFPKSIFH LKVPESSPIG SAIGRIRAVD PDFGQNAEIE YNIVPGDGGN LFDIVTDEDT QEGVIKLKKP LDFETKKAYT FKVEASNLHL DHRFHSAGPF KDTATVKISV LDVDEPPVFS KPLYTMEVYE DTPVGTIIGA VTAQDLDVGS SAVRYFIDWK SDGDSYFTID GNEGTIATNE LLDRESTAQY NFSIIASKVS NPLLTSKVNI LINVLDVNEF PPEISVPYET AVCENAKPGQ IIQIVSAADR DLSPAGQQFS FRLSPEAAIK PNFTVRDFRN NTAGIETRRN GYSRRQQELY FLPVVIEDSS YPVQSSTNTM TIRVCRCDSD GTILSCNVEA
Greater than 90% as determined by SDS-PAGE.
1010
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers