missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cadherin-12 Protein
A DNA sequence encoding the Homo sapiens (Human) Cadherin-12, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001304156.1 |
---|---|
Gene ID (Entrez) | 1010 |
Name | Cadherin-12 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026311
|
enQuireBio™
QP8756-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046311
|
enQuireBio™
QP8756-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036311
|
enQuireBio™
QP8756-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056311
|
enQuireBio™
QP8756-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066311
|
enQuireBio™
QP8756-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076311
|
enQuireBio™
QP8756-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001304156.1 | |
Cadherin-12 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1010 | |
Research Use Only | |
CDH12 | |
Recombinant Protein | |
E. coli | |
WVWNQFFVLE EYVGSEPQYV GKLHSDLDKG EGTVKYTLSG DGAGTVFTID ETTGDIHAIR SLDREEKPFY TLRAQAVDIE TRKPLEPESE FIIKVQDIND NEPKFLDGPY VATVPEMSPV GAYVLQVKAT DADDPTYGNS ARVVYSILQG QPYFSIDPKT GVIRTALPNM DREVKEQYQV LIQAKDMGGQ LGGLAGTTIV NITLTDVNDN PPRFPKSIFH LKVPESSPIG SAIGRIRAVD PDFGQNAEIE YNIVPGDGGN LFDIVTDEDT QEGVIKLKKP LDFETKKAYT FKVEASNLHL DHRFHSAGPF KDTATVKISV LDVDEPPVFS KPLYTMEVYE DTPVGTIIGA VTAQDLDVGS SAVRYFIDWK SDGDSYFTID GNEGTIATNE LLDRESTAQY NFSIIASKVS NPLLTSKVNI LINVLDVNEF PPEISVPYET AVCENAKPGQ IIQIVSAADR DLSPAGQQFS FRLSPEAAIK PNFTVRDFRN NTAGIETRRN GYSRRQQELY FLPVVIEDSS YPVQSSTNTM TIRVCRCDSD GTILSCNVEA | |
Greater than 90% as determined by SDS-PAGE. |