missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL21 / 6Ckine Protein
Recombinant Protein
Brand: enQuireBio™ QP5229-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002980.1 | |
CCL21 / 6Ckine Protein | |
Research Use Only | |
CCL21 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6366 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SDGGAQDCCL KYSQRKIPAK VVRSYRKQEP SLGCSIPAIL FLPRKRSQAE LCADPKELWV QQLMQHLDKT PSPQKPAQGC RKDRGASKTG KKGKGSKGCK RTERSQTPKG P | |
Greater than 95% as determined by SDS-PAGE. |