missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCNB2 Protein

A cDNA sequence encoding the CCNB2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11295-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956748

  • 203.00 € / 10µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

9133
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMALLRR PTVSSDLENI DTGVNSKVKS HVTIRRTVLE EIGNRVTTRA AQVAKKAQNT KVPVQPTKTT NVNKQLKPTA SVKPVQMEKL APKGPSPTPE DVSMKEENLC QAFSDALLCK IEDIDNEDWE NPQLCSDYVK DIYQYLRQLE VLQSINPHFL DGRDINGRMR AILVDWLVQV HSKFRLLQET LYMCVGIMDR FLQVQPVSRK KLQLVGITAL LLASKYEEMF SPNIEDFVYI TDNAYTSSQI REMETLILKE LKFELGRPLP LHFLRRASKA GEVDVEQHTL AKYLMELTLI DYDMVHYHPS KVAAAASCLS QKVLGQGKWN LKQQYYTGYT ENEVLEVMQH MAKNVVKVNE NLTKFIAIKN KYASSKLLKI SMIPQLNSKA VKDLASPLIG RSc
Greater than 90% as determined by SDS-PAGE.
CCNB2 Protein
Research Use Only
CCNB2
Human
His
The CCNB2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers