missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD40 Protein

A cDNA sequence encoding the CD40 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11333-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956898

  • 203.00 € / 10µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

958
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEPPTACR EKQYLINSQC CSLCQPGQKL VSDCTEFTET ECLPCGESEF LDTWNRETHC HQHKYCDPNL GLRVQQKGTS ETDTICTCEE GWHCTSEACE SCVLHRSCSP GFGVKQIATG VSDTICEPCP VGFFSNVSSA FEKCHPWTSC ETKDLVVQQA GTNKTDVVCG PQDRLR
Greater than 90.0% as determined by SDS-PAGE.
CD40 Protein
Research Use Only
CD40
Human
His
CD40 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers