missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human cDNA FLJ56323 Protein

A DNA sequence encoding the Homo sapiens (Human) cDNA FLJ56323, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8731-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046191

  • 2285.00 € / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002403.2
cDNA FLJ56323 Protein
Research Use Only
MGMT
Recombinant Protein
E. coli
QPAPLERFAS RRPQVLAVRT VCDLVLGKMD KDCEMKRTTL DSPLGKLELS GCEQGLHEIK LLGKGTSAAD AVEVPAPAAV LGGPEPLMQC TAWLNAYFHQ PEAIEEFPVP ALHHPVFQQE SFTRQVLWKL LKVVKFGEVI SYQQLAALAG NPKAARAVGG AMRGNPVPIL IPCHRVVCSS GAVGNYSGGL AVKEWLLAHE GHRLGKPGLG GSSGLAGAWL KGAGATSGSP PAGRN
Greater than 85.0% as determined by SDS-PAGE.
4255
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers