missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CENPA Protein

A cDNA sequence encoding the CENPA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11387-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927108

  • 5655.00 € / 1mg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CENPA Protein
Research Use Only
Recombinant Protein
Baculovirus
MGPRRRSRKP EAPRRRSPSP TPTPGPSRRG PSLGASSHQH SRRRQGWLKE IRKLQKSTHL LIRKLPFSRL AREICVKFTR GVDFNWQAQA LLALQEAAEA FLVHLFEDAY LLTLHAGRVT LFPKDVQLAR RIRGLEEGLG
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CENPA is supplied in 20mM HEPES buffer pH 8.0, 100mM sodium chloride, and 6M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers