missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Charged multivesicular body protein 4b Protein

A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 4b, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8215-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043331

  • 3260.00 € / 1mg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_789782.1
Charged multivesicular body protein 4b Protein
Research Use Only
CHMP4B
Recombinant Protein
E. coli
SVFGKLFGAG GGKAGKGGPT PQEAIQRLRD TEEMLSKKQE FLEKKIEQEL TAAKKHGTKN KRAALQALKR KKRYEKQLAQ IDGTLSTIEF QREALENANT NTEVLKNMGY AAKAMKAAHD NMDIDKVDEL MQDIADQQEL AEEISTAISK PVGFGEEFDE DELMAELEEL EQEELDKNLL EISGPETVPL PNVPSIALPS KPAKKKEEED DDMKELENWA GSM
Greater than 80% as determined by SDS-PAGE.
128866
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers