missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Charged multivesicular body protein 4b Protein
A DNA sequence encoding the Homo sapiens (Human) Charged multivesicular body protein 4b, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_789782.1 |
---|---|
Gene ID (Entrez) | 128866 |
Name | Charged multivesicular body protein 4b Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023331
|
enQuireBio™
QP8215-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043331
|
enQuireBio™
QP8215-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033331
|
enQuireBio™
QP8215-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053331
|
enQuireBio™
QP8215-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063331
|
enQuireBio™
QP8215-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073331
|
enQuireBio™
QP8215-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_789782.1 | |
Charged multivesicular body protein 4b Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
128866 | |
Research Use Only | |
CHMP4B | |
Recombinant Protein | |
E. coli | |
SVFGKLFGAG GGKAGKGGPT PQEAIQRLRD TEEMLSKKQE FLEKKIEQEL TAAKKHGTKN KRAALQALKR KKRYEKQLAQ IDGTLSTIEF QREALENANT NTEVLKNMGY AAKAMKAAHD NMDIDKVDEL MQDIADQQEL AEEISTAISK PVGFGEEFDE DELMAELEEL EQEELDKNLL EISGPETVPL PNVPSIALPS KPAKKKEEED DDMKELENWA GSM | |
Greater than 80% as determined by SDS-PAGE. |