missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COX5B Protein
A cDNA sequence encoding the COX5B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11492-10ug
Additional Details : Weight : 0.01000kg
Specifications
1329 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
COX5B | |
Human | |
His | |
COX5B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 20% glycerol and 1mM DTT. |
COX5B Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSASGGGVP TDEEQATGLE REIMLAAKKG LDPYNVLAPK GASGTREDPN LVPSISNKRI VGCICEEDNT SVVWFWLHKG EAQRCPRCGA HYKLVPQQLA H |