missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CST7 Protein
A cDNA sequence encoding the CST7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11546-10ug
Additional Details : Weight : 0.01000kg
Specifications
CST7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GPSPDTCSQD LNSRVKPGFP KTIKTNDPGV LQAARYSVEK FNNCTNDMFL FKESRITRAL VQIVKGLKYM LEVEIGRTTC KKNQHLRLDD CDFQTNHTLK QTLSCYSEVW VVPWLQHFEV PVLRCHHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CST7 solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4). |