missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CST7 Protein
A cDNA sequence encoding the CST7 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | CST7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927598
|
enQuireBio™
QP11546-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907598
|
enQuireBio™
QP11546-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917598
|
enQuireBio™
QP11546-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CST7 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CST7 solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
GPSPDTCSQD LNSRVKPGFP KTIKTNDPGV LQAARYSVEK FNNCTNDMFL FKESRITRAL VQIVKGLKYM LEVEIGRTTC KKNQHLRLDD CDFQTNHTLK QTLSCYSEVW VVPWLQHFEV PVLRCHHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |