missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTSL Protein
A cDNA sequence encoding the CTSL was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11561-1mg
Additional Details : Weight : 0.01000kg
Specifications
1514 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTLTFDHS LEAQWTKWKA MHNRLYGMNE EGWRRAVWEK NMKMIELHNQ EYREGKHSFT MAMNAFGDMT SEEFRQVMNG FQNRKPRKGK VFQEPLFYEA PRSVDWREKG YVTPVKNQGQ CGSCWAFSAT GALEGQMFRK TGRLISLSEQ NLVDCSGPQG NEGCNGGLMD YAFQYVQDNG GLDSEESYPY EATEESCKYN PKYSVANDTG FVDIPKQEKA LMKAVATVGP ISVAIDAGHE SFLFYKEGIY FEPDCSSEDM DHGVLVVGYG FESTESDNNK YWLVKNSWGE EWGMGGYVKM AKDRRNHCGI ASAASYPTV | |
Greater than 90% as determined by SDS-PAGE. |
CTSL Protein | |
Research Use Only | |
CTSL | |
Human | |
His | |
CTSL protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |