missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTSL Protein
A cDNA sequence encoding the CTSL was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 1514 |
---|---|
Name | CTSL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CTSL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957648
|
enQuireBio™
QP11561-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947648
|
enQuireBio™
QP11561-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937648
|
enQuireBio™
QP11561-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1514 | |
Research Use Only | |
CTSL | |
Human | |
His | |
CTSL protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
CTSL Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTLTFDHS LEAQWTKWKA MHNRLYGMNE EGWRRAVWEK NMKMIELHNQ EYREGKHSFT MAMNAFGDMT SEEFRQVMNG FQNRKPRKGK VFQEPLFYEA PRSVDWREKG YVTPVKNQGQ CGSCWAFSAT GALEGQMFRK TGRLISLSEQ NLVDCSGPQG NEGCNGGLMD YAFQYVQDNG GLDSEESYPY EATEESCKYN PKYSVANDTG FVDIPKQEKA LMKAVATVGP ISVAIDAGHE SFLFYKEGIY FEPDCSSEDM DHGVLVVGYG FESTESDNNK YWLVKNSWGE EWGMGGYVKM AKDRRNHCGI ASAASYPTV | |
Greater than 90% as determined by SDS-PAGE. |