missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL3/GRO gamma/CINC-2/DCIP-1 Protein
Alle ansehen Bio Techne Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 μg
100 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
500µg
Beschreibung
Fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected human 293 cells is in a concentration range of 10-100 ng/mL.
Spezifikation
Spezifikation
| Accession Number | P19876, P19876, P19876 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 50 mM NaCl. |
| Gene ID (Entrez) | 2921 |
| Molecular Weight (g/mol) | 7.9 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CXCL3/GRO gamma/CINC-2/DCIP-1 as determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 3, CINC-2 alpha, CINC-2 alpha/CXCL3, CINC-2 beta, CINC-2 beta/CXCL3, CINC-2b, C-X-C motif chemokine 3, DCIP-1, DCIP-1/CXCL3, GRO3, GRO3 oncogene, GROg, GRO-gamma, GRO-gamma(1-73), Growth-regulated protein gamma, Macrophage inflammatory protein 2-beta, melanoma growth stimulatory activity gamma, MGSA gamma, MIP-2b, MIP2B, MIP2-beta, SCYB3 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur