missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EDN2 Protein
A cDNA sequence encoding the EDN2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11746-5mg
Additional Details : Weight : 0.01000kg
Specifications
EDN2 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MGSSHHHHHH SSGLVPRGSH MGSQAAATLE QPASSSHAQG THLRLRRCSC SSWLDKECVY FCHLDIIWVN TPEQTAPYGL GNPPRRRRRS LPRRCQCSSA RDPACATFCL RRPWTEAGAV PSRKSPADVF QTGKTGATTG ELLQRLRDIS TVKSLFAKRQ QEAMREPRST HSRWRKR | |
Greater than 85.0% as determined by SDS-PAGE. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
EDN2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |