missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF (Leu 21) Protein
A cDNA sequence encoding the EGF (Leu 21) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10583-1mg
Additional Details : Weight : 0.01000kg
Specifications
EGF (Leu 21) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |