missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF (Leu 21) Protein
A cDNA sequence encoding the EGF (Leu 21) was constructed and used to recombinantly synthesize the protein.
131.00 € - 1355.00 €
Specifications
Name | EGF (Leu 21) Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994318
|
enQuireBio™
QP10583-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974318
|
enQuireBio™
QP10583-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984318
|
enQuireBio™
QP10583-1MG |
1 mg |
1355.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
EGF (Leu 21) Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
Untagged | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with no additives. |