missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8402-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044411

  • 2285.00 € / 1mg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003312.3
Elongation factor Tu, mitochondrial Protein
Research Use Only
TUFM
Recombinant Protein
E. coli
EAKKTYVRDK PHVNVGTIGH VDHGKTTLTA AITKILAEGG GAKFKKYEEI DNAPEERARG ITINAAHVEY STAARHYAHT DCPGHADYVK NMITGTAPLD GCILVVAAND GPMPQTREHL LLARQIGVEH VVVYVNKADA VQDSEMVELV ELEIRELLTE FGYKGEETPV IVGSALCALE GRDPELGLKS VQKLLDAVDT YIPVPARDLE KPFLLPVEAV YSVPGRGTVV TGTLERGILK KGDEC
Greater than 90% as determined by SDS-PAGE.
7284
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers