missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_003312.3 |
---|---|
Gene ID (Entrez) | 7284 |
Name | Elongation factor Tu, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024411
|
enQuireBio™
QP8402-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044411
|
enQuireBio™
QP8402-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034411
|
enQuireBio™
QP8402-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054411
|
enQuireBio™
QP8402-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064411
|
enQuireBio™
QP8402-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074411
|
enQuireBio™
QP8402-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003312.3 | |
Elongation factor Tu, mitochondrial Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7284 | |
Research Use Only | |
TUFM | |
Recombinant Protein | |
E. coli | |
EAKKTYVRDK PHVNVGTIGH VDHGKTTLTA AITKILAEGG GAKFKKYEEI DNAPEERARG ITINAAHVEY STAARHYAHT DCPGHADYVK NMITGTAPLD GCILVVAAND GPMPQTREHL LLARQIGVEH VVVYVNKADA VQDSEMVELV ELEIRELLTE FGYKGEETPV IVGSALCALE GRDPELGLKS VQKLLDAVDT YIPVPARDLE KPFLLPVEAV YSVPGRGTVV TGTLERGILK KGDEC | |
Greater than 90% as determined by SDS-PAGE. |