missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human EMAP-II/AIMP1 Protein

Product Code. 18422502 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
This item is not returnable. View return policy

Product Code. 18422502

Brand: Novus Biologicals™ NBP2349545ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by the apoptotic effect using serum free human MCF-7 cells is less than 40 ng/mL, corresponding to a specific activity of >2.5 x 10^4 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12904, Q12904, Q12904
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 9255
Molecular Weight (g/mol) 18.2 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of EMAP-II/AIMP1 as determined by LAL method.
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Gene Symbol AIMP1
Reconstitution Recommended to be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1 % BSA to a concentration of 0.1-1.0 mg/mL
Research Category Biologically Active Proteins, Cytokine Research
Purity or Quality Grade >98% pure by SDS-PAGE and HPLC
Protein EMAP-II/AIMP1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.