missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endoplasmic reticulum resident protein 29 Protein
A DNA sequence encoding the Homo sapiens (Human) Endoplasmic reticulum resident protein 29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8504-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001029197.1 | |
Endoplasmic reticulum resident protein 29 Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10961 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
C12orf8 | |
Recombinant Protein | |
E. coli | |
PLDTVTFYKV IPKSKFVLVK FDTQYPYGEK QDEFKRLAEN SASSDDLLVA EVGISDYGDK LNMELSEKYK LDKESYPVFY LFRDGDFENP VPYTGAVKVG AIQRWLKGQG VYLGMPGCLP VYDALAGEFI RASGVEARQA LLKQGQDNLS SVKETQKKWA EQYLKIMGKI LDQGEDFPAS EMTRIARLIE KNKMSDGKKE ELQKSLNILT AF |