missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP9 Protein
A cDNA sequence encoding the FABP9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11838-5ug
Additional Details : Weight : 0.01000kg
Specifications
FABP9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMVEPFL GTWKLVSSEN FEDYMKELGV NFAARNMAGL VKPTVTISVD GKMMTIRTES SFQDTKISFK LGEEFDETTA DNRKVKSTIT LENGSMIHVQ KWLGKETTIK RKIVDEKMVV ECKMNNIVST RIYEKV | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FABP9 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |