missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF6 Protein
A cDNA sequence encoding the FGF6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11872-25ug
Additional Details : Weight : 0.01000kg
Specifications
FGF6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
FGF-6 protein was lyophilized from a 0.2microm filtered solution in 10mM sodium phosphate and 50mM sodium chloride pH 7.5. |