missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF6 Protein
A cDNA sequence encoding the FGF6 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | FGF6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988648
|
enQuireBio™
QP11872-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978648
|
enQuireBio™
QP11872-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968648
|
enQuireBio™
QP11872-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FGF6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
FGF-6 protein was lyophilized from a 0.2microm filtered solution in 10mM sodium phosphate and 50mM sodium chloride pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI | |
Greater than 95.0% as determined by SDS-PAGE. |