missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FOLR1 Protein

A cDNA sequence encoding the FOLR1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11896-HIS-20ug

 View more versions of this product

Product Code. 15908728

  • 203.00 € / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

FOLR1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSRIAWART ELLNVCMNAK HHKEKPGPED KLHEQCRPWR KNACCSTNTS QEAHKDVSYL YRFNWNHCGE MAPACKRHFI QDTCLYECSP NLGPWIQQVD QSWRKERVLN VPLCKEDCEQ WWEDCRTSYT CKSNWHKGWN WTSGFNKCAV GAACQPFHFY FPTPTVLCNE IWTHSYKVSN YSRGSGRCIQ MWFDPAQGNP NEEVARFYAA AMS
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FOLR1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers