missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOLR1 Protein
A cDNA sequence encoding the FOLR1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | FOLR1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15998718
|
enQuireBio™
QP11896-HIS-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928728
|
enQuireBio™
QP11896-HIS-EC-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15908728
|
enQuireBio™
QP11896-HIS-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918728
|
enQuireBio™
QP11896-HIS-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988718
|
enQuireBio™
QP11896-HIS-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FOLR1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FOLR1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRIAWART ELLNVCMNAK HHKEKPGPED KLHEQCRPWR KNACCSTNTS QEAHKDVSYL YRFNWNHCGE MAPACKRHFI QDTCLYECSP NLGPWIQQVD QSWRKERVLN VPLCKEDCEQ WWEDCRTSYT CKSNWHKGWN WTSGFNKCAV GAACQPFHFY FPTPTVLCNE IWTHSYKVSN YSRGSGRCIQ MWFDPAQGNP NEEVARFYAA AMS | |
Greater than 90.0% as determined by SDS-PAGE. |