missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOLR1 Protein
A cDNA sequence encoding the FOLR1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11896-HIS-1mg
Informations supplémentaires : Weight : 0.01000kg
Specifications
FOLR1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRIAWART ELLNVCMNAK HHKEKPGPED KLHEQCRPWR KNACCSTNTS QEAHKDVSYL YRFNWNHCGE MAPACKRHFI QDTCLYECSP NLGPWIQQVD QSWRKERVLN VPLCKEDCEQ WWEDCRTSYT CKSNWHKGWN WTSGFNKCAV GAACQPFHFY FPTPTVLCNE IWTHSYKVSN YSRGSGRCIQ MWFDPAQGNP NEEVARFYAA AMS | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FOLR1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |