missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF-8 / Myostatin / MSTN Protein

Recombinant Protein

Brand:  enQuireBio™ QP5265-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985799

  • 4325.00 € / 500µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005250.1
GDF-8 / Myostatin / MSTN Protein
Research Use Only
MSTN
Recombinant Protein
E. coli
MNENSEQKEN VEKEGLCNAC TWRQNTKSSR IEAIKIQILS KLRLETAPNI SKDVIRQLLP KAPPLRELID QYDVQRDDSS DGSLEDDDYH ATTETIITMP TESDFLMQVD GKPKCCFFKF SSKIQYNKVV KAQLWIYLRP VETPTTVFVQ ILRLIKPMKD GTRYTGIRSL KLDMNPGTGI WQSIDVKTVL QNWLKQPESN LGIEIKALDE NGHDLAVTFP GPGEDGLNPF LEVKVTDTPK RSRR
Greater than 95% as determined by SDS-PAGE.
2660
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Bioactivity of Myostatin Propeptide was confirmed by measuring inhibition of myostatin induced cytotoxicity in MCP-11 cells. Typical ED50 is around 1 ug/ml myostatin propeptide.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers